1,1,2-Trimethoxyethane ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methoxyacetaldehyde dimethyl Acetal

T0831-25ML T0831-100ML
TCIAT0831-25MLEA 805 DKK
TCIAT0831-25ML TCIAT0831-100ML
1,1,2-Trimethoxyethane ≥98.0% (by GC)
1,1,2-Trimethoxyethane
Formel: C₅H₁₂O₃
Molekylvægt: 120.1470 g/mol
Kogepunkt: 126°C
Flammepunkt: 23°C
MDL Number: MFCD00008486
CAS-nummer: 24332-20-5
EINECS: 246-175-9
UN nummer: 3271
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aaa3dcfaba7c77ccbfb1f38b62ba6c4a3c5e5a6f&vid=aaa3dcfaba7c77ccbfb1f38b62ba6c4a3c5e5a6f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR