Didodecyldimethylammonium bromide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D1974-25G D1974-500G D1974-100G
TCIAD1974-25GEA 493 DKK
TCIAD1974-25G TCIAD1974-500G TCIAD1974-100G
Didodecyldimethylammonium bromide ≥98.0% (by titrimetric analysis)
Didodecyldimethylammonium bromide
Formel: C₂₆H₅₆BrN
MDL Number: MFCD00041969
CAS-nummer: 3282-73-3
EINECS: 221-923-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=068ff02ccb90f92fadcb1b73b51bf7f22d2334aa&vid=068ff02ccb90f92fadcb1b73b51bf7f22d2334aa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 161.0 to 167.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR