Strontiumperoxid (∼12,3% O₂)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
46310.A1
46310.A7
46310.30
46310.A1EA
3230
DKK
46310.A1
46310.A7
46310.30
Strontiumperoxid (∼12,3% O₂)
Strontium peroxide
CAS No.: 1314-18-7
Molecular Formula: O2Sr
Formula Weight: 119.62
Physical Form: -100 Mesh Powder
Sensitivity: Moisture Sensitive
Appearance: Pale brown
Melting Point: 215° dec.
UN No.: 1509
MDL No.: MFCD00084686
Product Specifications are subject to amendment and may change over time.
Formel:
O₂Sr Molekylvægt: 119.62 g/mol Smeltepunkt: 215 °C (dec.) Massefylde: 1.6 g/cm³ (60 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00084686 CAS-nummer: 1314-18-7 EINECS: 215-224-6 UN nummer: 1509 ADR: 5.1 II Merck Index: 14,8847 |
Specifikationer Testresultater
Available Oxygen | 12.3% min |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...