4-Amino-N,N-dimethylbenzensulfonamid 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H33747.03
H33747.06
H33747.03EA
969
DKK
H33747.03
H33747.06
4-Amino-N,N-dimethylbenzensulfonamid 97%
4-Amino-N,N-dimethylbenzensulfonamid
Formel:
C₈H₁₂N₂O₂S Smeltepunkt: 168…170 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00031428 CAS-nummer: 1709-59-7 EINECS: 216-970-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...