4-Methylthiobenzamid

Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Methylbenzothioamide, Thio-p-toluamide

13229-250 13229-500 13229-1 13229-5
CAYM13229-250EA 363 DKK
CAYM13229-250 CAYM13229-500 CAYM13229-1 CAYM13229-5
4-Methylthiobenzamid
4-Methylthiobenzamid
Formel: C₈H₉NS
Molekylvægt: 151.23
MDL Number: MFCD00173750
CAS-nummer: 2362-62-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR