4-Amino-o-cresol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Amino-2-methylphenol, 4-Hydroxy-3-methylaniline
A1536.0005
A1536-25G
A1536-5G
TCIAA1536.0005EA
1470
DKK
TCIAA1536.0005
TCIAA1536-25G
TCIAA1536-5G
4-Amino-o-cresol ≥98.0% (by GC, titration analysis)
4-Amino-o-cresol
Formel:
C₇H₉NO Kogepunkt: 259 °C (1013 hPa) Smeltepunkt: 129 °C |
MDL Number:
MFCD00007878 CAS-nummer: 2835-96-3 EINECS: 220-619-1 |
|
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...