Volumetric flasks, class A, brown markings, USP-individual certificate
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
US258/WAC/G/5 US258/WAC/H/2 US258/WAC/I/2 US258/WAC/J/2
POULUS258/WAC/G/5EA 1130 DKK
POULUS258/WAC/G/5 POULUS258/WAC/H/2 POULUS258/WAC/I/2 POULUS258/WAC/J/2
Volumetric flasks, class A, brown markings, USP-individual certificate
Kolber Volumetriske kolber
Borosilicate glass, clear, with PE stopper, USP certified.
  • Calibrated to contain (TC, IN)
  • Mark and inscriptions in brown
  • Supplied with USP-individual certificate
Certificeringer: DIN EN ISO 1042