VWR®, Separating funnels, conical
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
532-0007EA 405 DKK
532-0007 532-0008 532-0009 532-0010 532-0011 532-0012 532-0013 532-0014 532-0015 532-0006 532-0017 532-0016
VWR®, Separating funnels, conical
Tragte Skilletragte
Borosilicate 3.3 glass, with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance