Beaker Cups, PP, Medicine
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41102001 41102002 41102003 41102004 41102005
HECH41102001EA 21.6 DKK
HECH41102001 HECH41102002 HECH41102003 HECH41102004 HECH41102005
Beaker Cups, PP, Medicine
Bægre
PP, translucent.
  • Good temperature resistance
  • Good chemical resistance
  • Available in different colours
Ideal for general laboratory use and a wide variety of other applications. With embossed scale.