Spray bottles, turn 'n' spray, LaboPlast®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0309-0010 0309-0002 0309-0005
215-2963EA 209 DKK
215-2963 215-2961 215-2962
Spray bottles, turn 'n' spray, LaboPlast®
Flasker Sprayflasker
LDPE, translucent, PP spray attachment, PE tubing, stainless steel weight.
  • Uninterrupted spraying, even when inverted
  • Adjustable nozzle, diameter 0,6 mm
  • Amount sprayed per stroke: 1,2 ml ±0,1
Spraying upside down is not possible with standard spray bottles. As soon as the suction tube is no longer immersed in liquid, it cannot spray. Not so with the overhead spray bottle Turn 'n' Spray. It features a special technology that allows you to spray from any position. That means it can spray practically and conveniently from any direction and reach hard toMore Product Information