Bottle-top dispensers, FORTUNA® POLYFIX®
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
101.07048 101.07038 101.07138
615-0006EA 2710 DKK
615-0006 615-0005 POUL101.07138
Bottle-top dispensers, FORTUNA® POLYFIX®
Flasketopdispenser
Dispensers, made of PP, suitable for chemicals and most liquids.
  • Visible dispensing procedure
  • Easy-to-use volume setting system
  • Valve block made of polypropylene with FIX adapter
With glass piston: Suitable for all chemicals (including solvents), that do not attack polypropylene and do not crystallize.
With amber glass and glass piston: Suitable for all light sensitive liquids that do not attack polypropylene.
With coated PTFE-piston: Suitable for acids and alkaline solutions that do not attack polypropylene.
Leveringsinformation: Supplied with PTFE intake tube (1) and PP adaptors (2).