VWR®, Incinerating dishes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
459-0233EA 324 DKK
459-0233 459-0232 459-0231 459-0230 459-0229 459-0228 459-0227
VWR®, Incinerating dishes
Vejebåde Forbrændingsbåde
Porcelain, glazed, produced according to DIN 12904.
  • Dishes can withstand temperatures up to 1000 °C
  • Stable with large flat unglazed base
  • Glazed porcelain has high resistance to acids and bases