VWR®, Combustion boats
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
459-0223EA 623 DKK
459-0223 459-0226 459-0224 459-0225
VWR®, Combustion boats
Vejebåde Forbrændingsbåde
Porcelain.
  • Glazed boats can withstand temperatures up to 1000 °C, unglazed up to 1350 °C
  • Eyelet to aid recovery from furnace
  • Glazed porcelain has high resistance to acids and bases