VWR®, Bulb pipettes, one mark, class AS
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-4133EA 196 DKK
612-4133 612-4129 612-4130 612-4132 612-4134 612-4135 612-4136 612-4137 612-4138 612-4131
VWR®, Bulb pipettes, one mark, class AS
Pipetter Kolbepipetter
Borosilicate 9.1 glass.
  • Calibrated to deliver (TD)
  • Ring mark and inscriptions in high contrast amber stain
  • Not available with individual certificate
Certificeringer: DIN EN ISO 648
Bestillingsoplysninger: Dated batch identification and batch certificate included.