VWR®, Mortars, without Pestles
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
410-0117EA 2900 DKK
410-0117 410-0108 410-0118 410-0116 410-0115 410-0114 410-0113 410-0112 410-0111 410-0110 410-0109
VWR®, Mortars, without Pestles
Mortere og pistiller Mortere
Porcelain, produced according to DIN 12906.
  • With spout
  • Glazed outside, grinding surface unglazed
  • Highly resistant to acids and alkalis