Laboratory freezers, MediLine, LGPv series
Leverandør: Liebherr
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LGPV1420 LGPV1430 LGPV6530 LGPV8420 LGPV8430
472-2389EA 78591 DKK
472-2389 471-0661 471-0662 472-0118 471-1017
Laboratory freezers, MediLine, LGPv series
Frysere Laboratorie-/frysere til almindelige formål
These are heavy duty MediLine models with forced air cooling. The controller features door and temperature alarms which can be customised to suit the end user. Controller can be locked. The refrigerant used is hydrocarbon. Units have 83 to 60 mm insulation and 2 castors with brakes.
  • Back-up battery for 7,5 days data logging and 72 hours battery back up for controller in case of mains failure
  • RS485 interface connection, volt-free contacts, pre-drilled access port and optional infrared key for data download
  • Forced air ventilation with hot gas defrost - for
More Product Information
Bestil nu
Specifikationer
Model LGPv 1420 LPGv1430 LPGv6530 LPGv 8420 LPGv 8430
Environmentally Preferable
Art.nr 472-2389 471-0661 471-0662 472-0118 471-1017
Air circulation ventileret ventileret ventileret ventileret ventileret
Display LCD touch screen LCD touch screen LCD touch screen LCD touch screen
Defrost Automatic Automatic Automatic Automatic
Alarm Audible and visual Audible and visual Audible and visual Audible and visual
Shelves/drawers 2×4 shelves (24 levels max.) 4 shelves 5 shelves 4 shelves
Door type 2 doors 2 doors 1 door 1 door 1 door
Door lock Yes Yes Yes Yes
Access ports 1 for PT100 probe (not supplied), Ø 8 mm 1 for PT100 probe (not supplied), Ø 8 mm 1 for PT100 probe (not supplied), Ø 8 mm 1 for PT100 probe (not supplied), Ø 8 mm 1 for PT100 probe (not supplied), Ø 8 mm
Weight 262 kg 220 kg 144 kg 197 kg 203 kg