Shelving systems, stem castor carts with open grid shelves, MetroMax Q®
Leverandør: Metro International
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Q536EG3 Q556EG3 Q566BG3 Q356EG3 Q336EG3 Q436EG3
METOQ536EG3EA 11000 DKK
METOQ536EG3 METOQ556EG3 METOQ566BG3 METOQ356EG3 METOQ336EG3 METOQ436EG3
Shelving systems, stem castor carts with open grid shelves, MetroMax Q®
Møbler Hylder
Corrosion proof shelves and posts with removable polymer mats.
  • Load capacity: 408 kg (for models with PU castors); 272 kg (for models with resilient rubber castors)
Standard unit consist of 5 shelves (4 open grid, 1 bottom solid shelf), posts, wedge connectors and choice of PU or resilient rubber castors.