Askina® Elast fine Fixation Bandage
Leverandør: B. Braun
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9046089 9047085
BRAU9046089EA 214 DKK
BRAU9046089 BRAU9047085
Askina® Elast fine Fixation Bandage
Førstehjælpsudstyr Forbindinger
Elastic fixation bandage made from 54% polyamide and 46% viscose for securing wound dressings and splints and for light compression bandages.
  • Approx. 140% lengthways elasticity
  • Secure, long-lasting hold thanks to supple and elastic material
  • High stability thanks to woven edge
DIN 61634