DreamTaq™ PCR Master Mix (2X)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
K1071 K1072
K1071EA 1080 DKK
K1071 K1072
DreamTaq™ PCR Master Mix (2X)
Nukleinsyrereagenser End-point PCR Enzymes and Kits
DreamTaq™ PCR Master Mix (2X) is a ready to use solution containing DreamTaq™ DNA polymerase, optimised DreamTaq™ buffer, MgCl₂ and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR set-up. DreamTaq™ DNA polymerase is an enhanced Taq polymerase optimised for high throughput PCR applications. It ensures robust amplification of PCR products up to 6 kb from genomic DNA and up to 20 kb from viral DNA.
  • Convenient, ready to use
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Robust amplification of difficult templates
Leveringsinformation: Supplied with nuclease-free water.