Maxima™ Reverse Transcriptase
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EP0742 EP0743 EP0741
EP0742EA 2050 DKK
EP0742 EP0743 EP0741
Maxima™ Reverse Transcriptase
Nukleinsyrereagenser RT-PCR and cDNA Reagents
Maxima™ Reverse Transcriptase is an advanced enzyme derived by in vitro evolution of M-MuLV RT for RT-qPCR application. The enzyme features high thermostability, robustness and increased synthesis rates in RT-qPCR. The Maxima™ Reverse Transcriptase is capable of cDNA synthesis at elevated temperatures (50 to 60 °C) and completion of the reverse transcription reaction in 15 to 30 minutes. The use of this enzyme in combination with Maxima™ qPCR Master Mixes ensures high specificity, sensitivity and wide linear range of two step RT-qPCR. Applications include first strand cDNA synthesis, RT-PCR, and RT-qPCR.
  • Thermostable – 90% active after incubation at 50 °C for 60 minutes in a reaction mixture
  • Active up to 60 °C
  • High yields of full-length cDNA as long as 20 kb
  • Efficient – complete cDNA synthesis in 15 to 30 minutes
  • Incorporates modified nucleotides
  • Developed through in vitro evolution
Leveringsinformation: Supplied with 5X RT Buffer.