Volumetric spoons, disposable, SteriWare®
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8640H-20S 8640H-01S 8640H-05S 8640H-0123S 8640H-0246S 8640H-10S 8640H-10 8640H-01 8640H-20 8640H-05
SAMP8640H-20SEA 1270 DKK
SAMP8640H-20S SAMP8640H-01S SAMP8640H-05S SAMP8640H-0123S SAMP8640H-0246S SAMP8640H-10S SAMP8640H-10 SAMP8640H-01 SAMP8640H-20 SAMP8640H-05
Volumetric spoons, disposable, SteriWare®
Skeer
These volumetric spoons allow small volumes to be measured accurately. It made of HDPE and are designed to be used straight from the bag.
  • Moulded and packed in a cleanroom
  • BSE/TSE free
  • Full Batch Traceability
  • Fully validated sterilisation procedure. SAL 10⁻⁶
  • Available in a range of sizes from 1 to 20 ml
Suitable for food contact: FDA & EU 10/2011 conforming materials.
Emballering: Individually bagged.