Cleanroom bond paper, BioClean ISO Leaf™
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BICP-A4W80 BICP-A4P80 BICP-A4Y80 BICP-A4B80 BICP-A4G80
NITRBICP-A4W80EA 266 DKK
NITRBICP-A4W80 NITRBICP-A4P80 NITRBICP-A4Y80 NITRBICP-A4B80 NITRBICP-A4G80
Cleanroom bond paper, BioClean ISO Leaf™
Papir
Smooth paper surface, bagged in ISO Class 4/Class 10 cleanroom, individually lot numbered.
  • Latex free
  • Heat resistant
  • Contains no heavy metals
  • Low particle generation and ion levels
Emballering: 250 sheets per sealed PE bag; 10 PE bags per carton (2500 sheets).