Analogue stopwatches, split-second addition timer
Leverandør: HANHART
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
135.0201-0S 135.0301-0S 135.0101-0S 135.0401-0S
HANH135.0201-0SEA 6250 DKK
HANH135.0201-0S 609-0146 609-1129 609-1132
Analogue stopwatches, split-second addition timer
Alarmure Analoge stopure
Addition timer for time measurement with interrupt option, also drag indicator for split-time measurement.
  • Chrome plated brass casing, diamond cut
  • Lever, 7 jewels
  • Shock resistant, additional dust and water protection