Volumetric flasks, class A, blue markings
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.5124502F 1.5125102F 1.5125502F 1.5125902F 1.5124602F 1.5123702F 1.5124604F 1.5125104F 1.5124104F 1.5124304F 1.5125304F 1.5125504F
612-0543EA 137 DKK
612-0543 POUL1.5125102F POUL1.5125502F POUL1.5125902F 612-0555 POUL1.5123702F POUL1.5124604F POUL1.5125104F POUL1.5124104F POUL1.5124304F POUL1.5125304F POUL1.5125504F
Volumetric flasks, class A, blue markings
Kolber Volumetriske kolber
Borosilicate glass 3.3 with PP stopper.
  • Calibrated to contain (TC, In)
  • With ground ring mark
  • Conformity approved
Certificeringer: DIN EN ISO 1042