IV cannulae, Introcan Safety®
Leverandør: B. Braun
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4254511-01 4254554-01 4251679-01 4251644-01 4251614-01
BRAU4254511-01EA 1630 DKK
BRAU4254511-01 BRAU4254554-01 BRAU4251679-01 BRAU4251644-01 BRAU4251614-01
IV cannulae, Introcan Safety®
Kanyler
Introcan Safety passive shielded cannulae with self-activating protection against needlestick injuries and related infections.
  • Self-activating clip technology – No user activation required
  • No risk of inadvertent activation
  • Same puncture technique as with convential IV catheters