Dermatome scalpel blades, sterile
Leverandør: SWANN MORTON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9943 9944 9945
SWAN9943EA 1300 DKK
SWAN9943 SWAN9944 SWAN9945
Dermatome scalpel blades, sterile
Skalpeller Skalpelblade
These Dermatome blades are fitted into a handheld instrument which resembles an electric razor. The blade oscillates and the movement back and forth will "evenly" skim off the surface layers of skin. Electric Dermatomes are preferable for cutting thinner and longer strips of skin with a more homogeneous thickness.
  • Compatible with leading brands of Dermatome knives
  • Available in three sizes: 25 mm single segment, a 50 mm two segmental and a 75 mm three segmental blade
The Dermatome range is used in the surgical removal of damaged tissue and harvesting of "borrowed" tissue from a donor site during cosmetic procedures, reconstructive surgeries and Aesthetic treatments such as dermaplaning.
Emballering: Individually wrapped in foil packets and boxed in units of 10 pieces.