Hand shovels for industry
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8300-1003 8300-1001
BURK8300-1003EA 629 DKK
BURK8300-1003 BURK8300-1001
Hand shovels for industry
Spader
With anti-static additive, ideal for robust use in chemistry and industry. Polypropylene has a high resistance to chemicals and temperatures.
  • Anti-static
  • Unbreakable
  • Easy to clean
  • Light-weight
  • Autoclavable at 121 °C