Spatulas, Micro, Disposable, SteriWare®
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8015A-170 8015A-170S 8015V-170S 8015H-170 8015N-170 8015N-170S 8015NR-170S 8015H-170S 8015V-170
231-0488EA 1220 DKK
231-0488 231-0489 SAMP8015V-170S SAMP8015H-170 SAMP8015N-170 SAMP8015N-170S SAMP8015NR-170S SAMP8015H-170S SAMP8015V-170
Spatulas, Micro, Disposable, SteriWare®
Spatler
PS or HDPE, sterile or non sterile. These disposable micro spatulas are ideal for sampling small volumes.
  • Moulded and packed in a cleanroom
  • Suitable for food contact
  • Available sterile (gamma irradiated)
  • Individually packed
The narrow sampling blade enables samples to be taken from very small containers such as vials.