Sampler for Liquids, TeleScoop
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5065S-4500 5065S-3000 1035P-100 5065S-1500
SAMP5065S-4500EA 17750 DKK
SAMP5065S-4500 SAMP5065S-3000 SAMP1035P-100 SAMP5065S-1500
Sampler for Liquids, TeleScoop
Prøvetagere Væskeprøvetagere
These telescopic handles can be used when sampling from overhead gantries or difficult to reach places.
  • Strong telescopic handle
  • Range of volumes and styles of collecting vessels
Suitable for taking samples from liquids and suspensions.