Capillary columns, Elite-5 Amine for autosystem
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9316673 N9316676 N9316677 N9316680 N9316681 N9316682
PERCN9316673EA 21670 DKK
PERCN9316673 PERCN9316676 PERCN9316677 PERCN9316680 PERCN9316681 PERCN9316682
Capillary columns, Elite-5 Amine for autosystem
Kromatografikolonner
Elite-5 Amine capillary columns are ideal for the analysis of amines and other basic compounds.