Disposable bailers
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5040A-1000 5040W-1000
SAMP5040A-1000EA 3720 DKK
SAMP5040A-1000 SAMP5040W-1000
Disposable bailers
Prøvetagere Prøvetagere til engangsbrug
Disposable bailers designed for sampling from wells or storage silos.
Emballering: Individually bagged.
Advarsel: Not suitable for high density liquids.