Capillary columns, Elite-VMS for autosystem
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N9316650 N9316651 N9316652 N9316653 N9316654 N9316655
PERCN9316650EA 12580 DKK
PERCN9316650 PERCN9316651 PERCN9316652 PERCN9316653 PERCN9316654 PERCN9316655
Capillary columns, Elite-VMS for autosystem
Kromatografikolonner
Elite-VMS Pesticides capillary columns are specially designed for the analysis of volatile organic pollutants.