Bulb pipette, with piston, FORTUNA®
Leverandør: Poulten & Graf
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.2822502 1.2823102 1.2823602 1.2824502 1.2822504 1.2824104 1.2824504
POUL1.2822502EA 222 DKK
POUL1.2822502 POUL1.2823102 POUL1.2823602 POUL1.2824502 612-1259 612-1260 612-1261
Bulb pipette, with piston, FORTUNA®
Pipetter Kolbepipetter
Made of clear glass.
  • Safety pipette with suction piston
  • Blue or amber graduations
  • Calibrated cylinders and tolerance indications
  • Without certificate