Chromatographic columns with discs
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2.132.005 2.132.008 2.132.001 2.132.014
WITG2.132.005EA 1110 DKK
WITG2.132.005 WITG2.132.008 WITG2.132.001 WITG2.132.014
Chromatographic columns with discs
Kromatografikolonner
These borosilicate glass chromatography columns have fused-in sintered glass discs, ST-stopcock, with solid plug and screw-thread retaining nut.