Carboys, narrow neck, with screw cap and stopcock
Leverandør: Kautex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
130425-10/STRU 130425-5/STRUK
KAUT130425-10/STRUEA 653 DKK
KAUT130425-10/STRU KAUT130425-5/STRUK
Carboys, narrow neck, with screw cap and stopcock
Flasker Flasker, dunke
HDPE, natural colour.
  • Good chemical resistance and good thermal resistance from −40 to +110 °C
  • Clearly legible, blue litre graduations
Ideal for storing, transporting and decanting liquids. Also recommended for use as settling bottles. With one plastic-coated metal handle (5 and 10 litres) or two plastic handles (25 and 60 litres).