Faecal containers with screw cap
Leverandør: LP ITALIANA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
109247 109447 109147 109148 109048 109047 109448 109248
LPIT109247EA 2200 DKK
LPIT109247 LPIT109447 LPIT109147 LPIT109148 LPIT109048 LPIT109047 LPIT109448 LPIT109248
Faecal containers with screw cap
Kliniske diagnose systemer Clinical Sample Collection/Preparation Faeces Sample Collection
Containers made of PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
With or without spoon and label.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904