Condensers, Graham
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.734.250 1.737.250 1.738.250 1.732.160 1.733.250 1.733.400 1.734.400
WITG1.734.250EA 1150 DKK
WITG1.734.250 WITG1.737.250 WITG1.738.250 WITG1.732.160 WITG1.733.250 WITG1.733.400 WITG1.734.400
Condensers, Graham
Svaler
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Coil type, Graham pattern, suitable for either reflux or distillation work. Water connection with glass olives.