Overflow volumetric flasks, class B
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42115042 42115006 42115048 42115012 42115078 42115024 42115030 42115066 42115072 42115018
HECH42115042EA 326 DKK
HECH42115042 HECH42115006 HECH42115048 HECH42115012 HECH42115078 HECH42115024 HECH42115030 HECH42115066 HECH42115072 HECH42115018
Overflow volumetric flasks, class B
Kolber Volumetriske kolber
DURAN® borosilicate glass 3.3.
  • Acid-proof and alkali-resistant markings
  • Very good temperature resistance
  • Very good chemical resistance
For water analysis.