Measuring cylinders, tall form, class B, soda glass
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42200030 42200048 42200060 42200036 42200042 42200024 42200066 42200018 42200006 42200012
612-9299EA 90.1 DKK
612-9299 612-9449 HECH42200060 HECH42200036 HECH42200042 HECH42200024 HECH42200066 HECH42200018 HECH42200006 HECH42200012
Measuring cylinders, tall form, class B, soda glass
Cylindere Målecylindre
Soda glass, tall form, class B.
  • Calibrated to contain (marked 'In')
  • Round base, spout
  • Fused amber short line graduation
Certificeringer: DIN EN ISO 4788