Accessories for beakers, low form, PTFE, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H927-14 H927-06 H927-08 H927-13 H927-11 H927-09 H927-07 H927-05 H927-04 H927-03 H927-15
BOHLH927-14EA 1490 DKK
BOHLH927-14 BOHLH927-06 BOHLH927-08 BOHLH927-13 BOHLH927-11 BOHLH927-09 BOHLH927-07 BOHLH927-05 BOHLH927-04 BOHLH927-03 BOHLH927-15
Accessories for beakers, low form, PTFE, BOLA
Bægre
PTFE, white.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +250 °C
Lid with centering shoulder.