Racks for centrifuge tubes, PP
Leverandør: GLW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G23 G26 G18
391-8848EA 246 DKK
391-8848 391-8849 GLWAG18
Racks for centrifuge tubes, PP
Stativer Tube Racks
  • Unique screw assembly provides superior durability and assured stability
  • Space efficient designed that is for use in the safety cabinet
  • Use in freezer down to −90 °C, autoclavable up to +121 °C