Cryoboxes, Cardboard
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
820003 820100 820081 820064 820049 820016 820002 5960 820025 820109
HERA820003EA 53.1 DKK
HERA820003 HERA820100 HERA820081 HERA820064 HERA820049 HERA820016 HERA820002 HERA5960 HERA820025 HERA820109
Cryoboxes, Cardboard
Bokse Æsker til Kryorør
Cryo resistant cardboard.
  • Use with cardboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.