Acridine orange (hemizinc salt)

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

0249.1 0249.2
ROTH0249.1EA 0 DKK
ROTH0249.1 ROTH0249.2
Acridine orange (hemizinc salt)
Acridine orange (hemizinc salt)
Formel: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Molekylvægt: 369.96 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081043
CAS-nummer: 10127-02-3
EINECS: 233-353-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR