Acridine orange (hemizinc salt) ∼55% (dye content), pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

300910250 300911000
ACRO300910250EA 369 DKK
ACRO300910250 ACRO300911000
Acridine orange (hemizinc salt) ∼55% (dye content), pure
Acridine orange (hemizinc salt)
Formel: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Molekylvægt: 369.96 g/mol
Smeltepunkt: 165 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081043
CAS-nummer: 10127-02-3
EINECS: 233-353-6
UN nummer: 3143
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance Red-brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR