Acriflavine neutral 13,3-15,8% available chlorine
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base
211400250 211401000
ACRO211400250EA 771 DKK
ACRO211400250 ACRO211401000
Acriflavine neutral 13,3-15,8% available chlorine
Acriflavine
Formel: C₂₇H₂₅ClN₆
Molekylvægt: 468.9806 g/mol
Smeltepunkt: 179…181 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064307
CAS-nummer: 8048-52-0
Bestil nu
Specifikationer Testresultater
Appearance Orange to red-brown Powder
Infrared spectrum Conforms
Titration Argentometric 13.3 to 15.8 % (Chlorine) on dried substance
Loss on drying ≤8 % (105°C, 2 h)