Tetrachlorobis(tetrahydrofuran)zirconium(IV)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Tetrachlorobis(tetrahydrofuran)zirconium

39550.04 39550.09
39550.04EA 596 DKK
39550.04 39550.09
Tetrachlorobis(tetrahydrofuran)zirconium(IV)
Tetrachlorobis(tetrahydrofuran)zirconium(IV)

CAS No.: 21959-01-3 Molecular Formula: C8H16Cl4O2Zr Formula Weight: 377.25 Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White to off-white Melting Point: 175-177° UN No.: 2925 MDL No.: MFCD00145367

Formel: C₄H₈Cl₄OZr
Smeltepunkt: 175…177 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00145367
CAS-nummer: 21959-01-3
UN nummer: 2925
ADR: 4.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR