4-Nitroquinoline-N-oxide 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Nitroquinoline 1-oxide, 4-Nitroquinoline N-oxide

203790010 203792500 203790050
ACRO203790010EA 692 DKK
ACRO203790010 ACRO203792500 ACRO203790050
4-Nitroquinoline-N-oxide 98%
4-Nitroquinoline-N-oxide
Formel: C₉H₆N₂O₃
Molekylvægt: 190.1555 g/mol
Smeltepunkt: 154…156 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006738
CAS-nummer: 56-57-5
EINECS: 200-281-1

Bestil nu

Specifikationer Testresultater

Appearance Ochre to brown Crystalline solid
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR