N-Methyl-bis-trifluoroacetamide, Supelco®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA
M-Clarity™ quality level = MQ100
65943-25ML
65943-5ML
65943-25MLEA
5890
DKK
65943-25ML
65943-5ML
N-Methyl-bis-trifluoroacetamide, Supelco®
N-Methylbistrifluoroacetamide
Formel:
C₅H₃F₆NO₂ |
MDL Number:
MFCD00000412 CAS-nummer: 685-27-8 EINECS: 211-680-5 UN nummer: 2920 ADR: 8.3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...