Methyl green zinc chloride salt

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: CI 42590, Ethyl Green, Methyl green zinc chloride double salt, Methyl Green

29295.01 29295.02
SERA29295.01EA 0 DKK
SERA29295.01 SERA29295.02
Methyl green zinc chloride salt
Methyl green zinc chloride salt
Formel: C₂₇H₃₅BrClN₃·xZnCl₂
Molekylvægt: 653.24 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00151094
CAS-nummer: 7114-03-6
EINECS: 230-415-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR