(+)-Dibenzoyl-D-tartaric acid 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid
302510250
302511000
302515000
ACRO302510250EA
316
DKK
ACRO302510250
ACRO302511000
ACRO302515000
(+)-Dibenzoyl-D-tartaric acid 98+%
(+)-Dibenzoyl-D-tartaric acid
Formel:
C₁₈H₁₄O₈ Smeltepunkt: 150…155 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00063222 CAS-nummer: 17026-42-5 EINECS: 241-097-1 |
Specifikationer Testresultater
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 150 to 155 °C |
HPLC | ≥98.0 % |
Specific optical rotation | +117° to +123° (20 °C, 589 nm) (c=5, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...